Assay Details
Target Gene Details
Entrez Gene ID: | 55236 |
Gene Name: | ubiquitin like modifier activating enzyme 6 |
Gene Aliases: |
E1-L2, MOP-4, UBE1L2 |
Location: |
Chr.4:67615761-67701171 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 26 - Exon 26 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UBA6 | NM_018227.5 | NP_060697.4 | ||
| XM_017008359.1 | XP_016863848.1 | |||
| AB014773.1 | BAB19785.1 | |||
| AK001670.1 | BAA91824.1 | |||
| AK094164.1 | ||||
| AK314371.1 | ||||
| AL832015.2 | CAD89908.1 | |||
| AL832458.2 | CAD89959.1 | |||
| AY359880.1 | AAQ63403.1 | |||
| BC126484.1 | ||||
| BC126486.1 | ||||
| EF623993.1 |
More Information
Set Membership: |
|

Genomic Map