Assay Details
Target Gene Details
Entrez Gene ID: | 617 |
Gene Name: | BCS1 homolog, ubiquinol-cytochrome c reductase complex chaperone |
Gene Aliases: |
BCS, BCS1, BJS, FLNMS, GRACILE, Hs.6719, MC3DN1, PTD, h-BCS, h-BCS1 |
Location: |
Chr.2:218659656-218663443 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
More Information
Set Membership: |
|

Genomic Map