Assay Details
Target Gene Details
Entrez Gene ID: | 152110 |
Gene Name: | NIMA related kinase 10 |
Gene Aliases: |
- |
Location: |
Chr.3:27107336-27369460 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 43 - Intron 44 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv880e214 | Chr.3:26983690 - 27243989 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map