Assay Details
Target Gene Details
Entrez Gene ID: | 153364 |
Gene Name: | metallo-beta-lactamase domain containing 2 |
Gene Aliases: |
- |
Location: |
Chr.5:90458203-90474768 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MBLAC2 | NM_203406.1 | 2 | 1239 | NP_981951.1 |
| AK128603.1 | 3 | 649 | ||
| AK289778.1 | 2 | 791 | ||
| BC038230.1 | 2 | 1239 | AAH38230.1 | |
| CR749512.1 | 1 | 1579 | CAH18329.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv518131 | Chr.5:90452325 - 90485777 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map