Assay Details
Target Gene Details
Entrez Gene ID: | 199990 |
Gene Name: | Fanconi anemia core complex associated protein 20 |
Gene Aliases: |
C1orf86, FP7162 |
Location: |
Chr.1:2184460-2212720 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAAP20 | NM_001146310.1 | NP_001139782.1 | ||
| NM_001256945.1 | NP_001243874.1 | |||
| NM_001282671.1 | 7 | 1102 | NP_001269600.1 | |
| NM_001282672.1 | 6 | 971 | NP_001269601.1 | |
| NM_001282673.1 | NP_001269602.1 | |||
| XM_017000554.1 | XP_016856043.1 | |||
| XM_017000555.1 | 1 | 1138 | XP_016856044.1 | |
| XM_017000556.1 | 2 | 550 | XP_016856045.1 | |
| AF469129.1 | 7 | 1068 | AAP97719.1 | |
| AK055593.1 | 6 | 971 | BAB70965.1 | |
| AK127994.1 | BAC87220.1 | |||
| BG825658.1 | ||||
| BU192678.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv482937 | Chr.1:10001 - 2368561 on Build GRCh38 | Loss |
|
| dgv49n54 | Chr.1:2151050 - 2289783 on Build GRCh38 | Loss |
|
| nsv951530 | Chr.1:2153662 - 2229961 on Build GRCh38 | Deletion |
|
| nsv834557 | Chr.1:2117631 - 2311104 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map