Assay Details
Target Gene Details
Entrez Gene ID: | 788 |
Gene Name: | solute carrier family 25 member 20 |
Gene Aliases: |
CAC, CACT |
Location: |
Chr.3:48856923-48898993 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC25A20 | NM_000387.5 | 9 | 1206 | NP_000378.1 |
| AK301744.1 | 10 | 1167 | ||
| BC001689.2 | 9 | 1078 | AAH01689.1 | |
| BQ018220.1 | ||||
| Y10319.1 | 9 | 1069 | CAA71367.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3893694 | Chr.3:48792101 - 48921349 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map