Assay Details
Target Gene Details
Entrez Gene ID: | 11018 |
Gene Name: | transmembrane p24 trafficking protein 1 |
Gene Aliases: |
IL1RL1LG, Il1rl1l, Tp24, p24g1 |
Location: |
Chr.19:10832067-10836307 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMED1 | NM_006858.3 | 3 | 522 | NP_006849.1 |
| NR_104015.1 | 3 | 361 | ||
| XM_006722631.3 | 3 | 352 | XP_006722694.1 | |
| BC002443.1 | 3 | 410 | AAH02443.1 | |
| BF794897.1 | 3 | 522 | ||
| HY012010.1 | 3 | 362 | ||
| U41804.1 | 3 | 493 | AAC50419.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1160584 | Chr.19:10618877 - 10892186 on Build GRCh38 | Deletion |
|
| nsv833748 | Chr.19:10757002 - 10925368 on Build GRCh38 | Loss |
|
| nsv2416 | Chr.19:10814223 - 10848907 on Build GRCh38 | Insertion |
|
| nsv833746 | Chr.19:10678751 - 10842756 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map