Assay Details
Target Gene Details
Entrez Gene ID: | 22 |
Gene Name: | ATP binding cassette subfamily B member 7 |
Gene Aliases: |
ABC7, ASAT, Atm1p, EST140535 |
Location: |
Chr.X:75053172-75156340 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ABCB7 | NM_001271696.1 | 12 | 1666 | NP_001258625.1 |
| NM_001271697.1 | 11 | 1546 | NP_001258626.1 | |
| NM_001271698.1 | 11 | 1588 | NP_001258627.1 | |
| NM_001271699.1 | 11 | 1549 | NP_001258628.1 | |
| NM_004299.4 | 12 | 1669 | NP_004290.2 | |
| AF038950.1 | 12 | 1606 | AAC39865.1 | |
| AF078777.1 | 12 | 1586 | AAD47141.1 | |
| AF133659.1 | 12 | 1598 | AAD33045.1 | |
| AK001418.1 | 12 | 1626 | ||
| AK294657.1 | 11 | 1530 | ||
| AK307414.1 | 11 | 1469 | ||
| BC006323.2 | 12 | 1602 | AAH06323.1 | |
| BT009918.1 | 12 | 1601 | AAP88920.1 | |
| BX537833.1 | 11 | 1486 | CAD97847.1 | |
| KU177872.1 | 12 | 1601 | ||
| KU177873.1 | ||||
| KU177874.1 | ||||
| KU177875.1 | ||||
| U66679.1 | 3 | 319 |
More Information
Set Membership: |
|

Genomic Map