Assay Details
Target Gene Details
Entrez Gene ID: | 8001 |
Gene Name: | glycine receptor alpha 3 |
Gene Aliases: |
- |
Location: |
Chr.4:174642011-174829568 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GLRA3 | NM_001042543.2 | 9 | 1884 | NP_001036008.1 |
| NM_006529.3 | 10 | 1929 | NP_006520.2 | |
| XM_011532268.2 | 6 | 1175 | XP_011530570.1 | |
| XM_017008630.1 | 11 | 1953 | XP_016864119.1 | |
| AF018157.1 | 4 | 692 | AAC39918.1 | |
| BC036086.1 | 10 | 1928 | AAH36086.1 | |
| U93917.1 | 9 | 1800 | AAC39917.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3603233 | Chr.4:174643234 - 174652749 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map