Assay Details
Target Gene Details
Entrez Gene ID: | 1183 |
Gene Name: | chloride voltage-gated channel 4 |
Gene Aliases: |
CLC4, ClC-4, ClC-4A |
Location: |
Chr.X:10156945-10237660 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CLCN4 | NM_001256944.1 | NP_001243873.1 | ||
| NM_001830.3 | NP_001821.2 | |||
| AB019432.1 | BAA77327.1 | |||
| AK225287.1 | ||||
| AK289564.1 | ||||
| BC130278.1 | ||||
| BP310547.1 | ||||
| X77197.1 | CAA54417.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2758861 | Chr.X:10083096 - 10263971 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map