Assay Details
Target Gene Details
Entrez Gene ID: | 2632 |
Gene Name: | glucan (1,4-alpha-), branching enzyme 1 |
Gene Aliases: |
APBD, GBE, GSD4 |
Location: |
Chr.3:81489699-81761799 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GBE1 | NM_000158.3 | 16 | 2982 | NP_000149.3 |
| AK125918.1 | 16 | 3345 | ||
| BC012098.1 | AAH12098.1 | |||
| BM509929.1 | 1 | 118 | ||
| BQ022720.1 | 1 | 137 | ||
| BQ632568.1 | ||||
| L07956.1 | 16 | 2789 | AAA58642.1 |
More Information
Set Membership: |
|

Genomic Map