Assay Details
Target Gene Details
Entrez Gene ID: | 9406 |
Gene Name: | zinc finger RANBP2-type containing 2 |
Gene Aliases: |
ZIS, ZIS1, ZIS2, ZNF265 |
Location: |
Chr.1:71063291-71081289 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZRANB2 | NM_005455.4 | NP_005446.2 | ||
| NM_203350.2 | NP_976225.1 | |||
| AB209568.1 | BAD92805.1 | |||
| AF065391.1 | AAD09746.1 | |||
| AF065392.1 | AAD09747.1 | |||
| AK023252.1 | ||||
| AK222858.1 | BAD96578.1 | |||
| AL136945.1 | CAB66879.1 | |||
| BC039814.1 | AAH39814.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv546472 | Chr.1:70967301 - 71075320 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map