Assay Details
Target Gene Details
Entrez Gene ID: | 10149 |
Gene Name: | adhesion G protein-coupled receptor G2 |
Gene Aliases: |
EDDM6, GPR64, HE6, TM7LN2 |
Location: |
Chr.X:18989307-19122956 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 23 - Intron 24 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv33517 | Chr.X:18661048 - 19128069 on Build GRCh38 | Loss |
|
| esv3576793 | Chr.X:18780019 - 19298577 on Build GRCh38 | Gain |
|
| esv2752317 | Chr.X:18691267 - 19292303 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map