Assay Details
Target Gene Details
Entrez Gene ID: | 9581 |
Gene Name: | prolyl endopeptidase-like |
Gene Aliases: |
- |
Location: |
Chr.2:44317607-44361862 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PREPL | NM_001042385.2 | 13 | 3919 | NP_001035844.1 |
| NM_001042386.2 | 13 | 3907 | NP_001035845.1 | |
| NM_001171603.1 | 15 | 4445 | NP_001165074.1 | |
| NM_001171606.1 | 15 | 4576 | NP_001165077.1 | |
| NM_001171613.1 | 14 | 4034 | NP_001165084.1 | |
| NM_001171617.1 | 14 | 4191 | NP_001165088.1 | |
| NM_006036.4 | 14 | 5030 | NP_006027.2 | |
| XM_011533198.1 | 15 | 4532 | XP_011531500.1 | |
| XM_011533200.1 | 15 | 4184 | XP_011531502.1 | |
| XM_011533202.1 | 14 | 3984 | XP_011531504.1 | |
| XM_017005384.1 | 15 | 5170 | XP_016860873.1 | |
| XM_017005385.1 | 15 | 4495 | XP_016860874.1 | |
| AB007896.1 | 14 | 3957 | ||
| BC151236.1 | 15 | 4509 | ||
| DQ023503.1 | 14 | 5030 | AAY89634.1 | |
| DQ023504.1 | 14 | 5030 | AAY89635.1 |
Target Gene Details
Entrez Gene ID: | 6519 |
Gene Name: | solute carrier family 3 member 1 |
Gene Aliases: |
ATR1, CSNU1, D2H, NBAT, RBAT |
Location: |
Chr.2:44275458-44320824 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC3A1 | NM_000341.3 | NP_000332.2 | ||
| AB033549.1 | BAB16841.1 | |||
| AK091932.1 | ||||
| AK092000.1 | ||||
| AK223146.1 | BAD96866.1 | |||
| AK289636.1 | ||||
| AK313157.1 | ||||
| BC022386.1 | AAH22386.1 | |||
| BC093624.1 | AAH93624.1 | |||
| BC093626.1 | AAH93626.1 | |||
| DQ023512.1 | AAY89643.1 | |||
| DQ023514.1 | AAY89645.1 | |||
| KU178335.1 | ||||
| KU178336.1 | ||||
| L11696.1 | AAA81778.1 | |||
| M95548.1 | AAA35500.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv3798n100 | Chr.2:44279994 - 44404126 on Build GRCh38 | Loss |
|
| nsv581692 | Chr.2:44278046 - 44322729 on Build GRCh38 | Loss |
|
| dgv3799n100 | Chr.2:44284605 - 44325240 on Build GRCh38 | Loss |
|
| dgv6838n54 | Chr.2:44297436 - 44341555 on Build GRCh38 | Loss |
|
| nsv1006405 | Chr.2:44013164 - 44709137 on Build GRCh38 | Loss |
|
| nsv1011655 | Chr.2:44278899 - 44344334 on Build GRCh38 | Loss |
|
| esv3892026 | Chr.2:44318437 - 44418772 on Build GRCh38 | Loss |
|
| nsv581691 | Chr.2:44276990 - 44339733 on Build GRCh38 | Gain |
|
| esv3590555 | Chr.2:44306026 - 44320888 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map