Assay Details
Target Gene Details
Entrez Gene ID: | 54107 |
Gene Name: | DNA polymerase epsilon 3, accessory subunit |
Gene Aliases: |
CHARAC17, CHRAC17, YBL1, p17 |
Location: |
Chr.9:113407235-113410749 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| POLE3 | NM_001278255.1 | 5 | 1375 | NP_001265184.1 |
| NM_017443.4 | 5 | 1405 | NP_059139.3 | |
| NR_027261.1 | 4 | 1539 | ||
| AF070640.1 | 1 | 726 | ||
| AK074629.1 | 4 | 1249 | BAC11099.1 | |
| AK074762.1 | 5 | 1328 | BAC11190.1 | |
| AK074782.1 | 4 | 1238 | BAC11206.1 | |
| AK092840.1 | 3 | 1533 | ||
| AK096050.1 | 4 | 1182 | ||
| BC003166.1 | 4 | 1228 | AAH03166.1 | |
| BC004170.2 | 4 | 1228 | AAH04170.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3621540 | Chr.9:113279513 - 113421563 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map