Assay Details
Target Gene Details
Entrez Gene ID: | 23076 |
Gene Name: | ribosomal RNA processing 1B |
Gene Aliases: |
KIAA0179, NNP1L, Nnp1, PPP1R136, RRP1 |
Location: |
Chr.21:43659551-43696079 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RRP1B | NM_015056.2 | NP_055871.1 | ||
| AK056263.1 | ||||
| BC014005.2 | AAH14005.1 | |||
| BC028386.2 | AAH28386.1 | |||
| D80001.1 | BAA11496.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv587700 | Chr.21:43588075 - 43675319 on Build GRCh38 | Loss |
|
| esv999311 | Chr.21:43667857 - 43677707 on Build GRCh38 | Deletion |
|
| nsv1066728 | Chr.21:43669996 - 43711842 on Build GRCh38 | Loss |
|
| nsv834106 | Chr.21:43572506 - 43689608 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map