Assay Details
Target Gene Details
Entrez Gene ID: | 729991 |
Gene Name: | BLOC-1 related complex subunit 8 |
Gene Aliases: |
MEF2BNB |
Location: |
Chr.19:19176904-19192591 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BORCS8 | NM_001145783.1 | 1 | 70 | NP_001139255.1 |
| NM_001145784.1 | 1 | 70 | NP_001139256.1 | |
| DA440390.1 | 1 | 70 |
Target Gene Details
Entrez Gene ID: | 4207 |
Gene Name: | BORCS8-MEF2B readthrough |
Gene Aliases: |
LOC729991-MEF2B, MEF2B, MEF2BNB-MEF2B |
Location: |
Chr.19:19145567-19192591 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BORCS8-MEF2B | NM_005919.3 | 1 | 70 | NP_005910.1 |
| NR_027307.1 | 1 | 70 | ||
| NR_027308.1 | 1 | 70 |
Target Gene Details
Entrez Gene ID: | 8625 |
Gene Name: | regulatory factor X associated ankyrin containing protein |
Gene Aliases: |
ANKRA1, BLS, F14150_1, RFX-B |
Location: |
Chr.19:19192199-19201869 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RFXANK | NM_001278727.1 | 1 | 324 | NP_001265656.1 |
| NM_003721.3 | 1 | 324 | NP_003712.1 | |
| NM_134440.2 | 1 | 324 | NP_604389.1 | |
| XM_005260136.4 | 1 | 288 | XP_005260193.1 | |
| AF094760.1 | 1 | 236 | AAC69883.1 | |
| BC114558.1 | 1 | 280 | AAI14559.1 | |
| BC114563.1 | 1 | 280 | AAI14564.1 | |
| BM682278.1 | 1 | 346 | ||
| CX871218.1 | 1 | 257 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833779 | Chr.19:19052683 - 19202282 on Build GRCh38 | Loss |
|
| nsv1151217 | Chr.19:18627369 - 19284914 on Build GRCh38 | Deletion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs73922821] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map