Assay Details
Target Gene Details
Entrez Gene ID: | 144100 |
Gene Name: | pleckstrin homology domain containing A7 |
Gene Aliases: |
- |
Location: |
Chr.11:16787153-17014423 on Build GRCh38 |
Assay Gene Location: | Within Exon 25 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PLEKHA7 | NM_175058.4 | 23 | 4334 | NP_778228.3 |
| XM_017017240.1 | XP_016872729.1 | |||
| XM_017017241.1 | 25 | 4545 | XP_016872730.1 | |
| XM_017017242.1 | 24 | 4425 | XP_016872731.1 | |
| XM_017017243.1 | 23 | 4344 | XP_016872732.1 | |
| AW135567.1 | ||||
| BC033239.1 | 14 | 3214 | AAH33239.1 | |
| BC041452.1 | 5 | 1672 | AAH41452.1 | |
| BC071599.1 | 23 | 4330 | AAH71599.1 | |
| BX648885.1 | 2 | 3117 |
More Information
Set Membership: |
|

Genomic Map