Assay Details
Target Gene Details
Entrez Gene ID: | 64121 |
Gene Name: | Ras related GTP binding C |
Gene Aliases: |
GTR2, RAGC, TIB929 |
Location: |
Chr.1:38838197-38859823 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RRAGC | NM_001271851.1 | 6 | 1025 | NP_001258780.1 |
| NM_022157.3 | 6 | 1127 | NP_071440.1 | |
| AF272035.1 | 6 | 955 | AAG32662.1 | |
| AF323609.1 | 6 | 950 | AAG45221.1 | |
| AK023373.1 | 2 | 155 | BAB14548.1 | |
| AK092808.1 | 1 | 2038 | ||
| AK298573.1 | 5 | 650 | ||
| AK298788.1 | 6 | 921 | ||
| AK315791.1 | 6 | 1052 | ||
| BC016668.1 | 6 | 972 | AAH16668.1 | |
| CA449149.1 | 2 | 473 | ||
| CX761032.1 |
More Information
Set Membership: |
|

Genomic Map