Assay Details
Target Gene Details
Entrez Gene ID: | 2960 |
Gene Name: | general transcription factor IIE subunit 1 |
Gene Aliases: |
FE, TF2E1, TFIIE-A |
Location: |
Chr.3:120742711-120783134 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GTF2E1 | NM_005513.2 | 2 | 487 | NP_005504.2 |
| XM_011512744.2 | 2 | 610 | XP_011511046.1 | |
| XM_011512745.2 | 2 | 558 | XP_011511047.1 | |
| AK223401.1 | 2 | 453 | BAD97121.1 | |
| BC005917.1 | 2 | 436 | ||
| DA498425.1 | 2 | 487 | ||
| S67859.1 | 2 | 436 | AAB20413.1 | |
| X63468.1 | 1 | 427 | CAA45068.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv4847n100 | Chr.3:120741894 - 120864319 on Build GRCh38 | Gain |
|
| nsv829699 | Chr.3:120589485 - 120765848 on Build GRCh38 | Loss |
|
| nsv829700 | Chr.3:120750344 - 120956916 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map