Assay Details
Target Gene Details
Entrez Gene ID: | 1787 |
Gene Name: | tRNA aspartic acid methyltransferase 1 |
Gene Aliases: |
DMNT2, DNMT2, MHSAIIP, PUMET, RNMT1 |
Location: |
Chr.10:17136785-17202071 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TRDMT1 | NM_001321006.1 | 9 | 5759 | NP_001307935.1 |
| NM_001321007.1 | 12 | 6132 | NP_001307936.1 | |
| NM_004412.6 | 11 | 5902 | NP_004403.1 | |
| XM_005252374.4 | XP_005252431.1 | |||
| XM_005252375.4 | 9 | 5759 | XP_005252432.1 | |
| XM_006717388.3 | 11 | 5876 | XP_006717451.1 | |
| XM_011519343.2 | 10 | 5831 | XP_011517645.1 | |
| XM_011519344.2 | 10 | 5820 | XP_011517646.1 | |
| XM_011519345.2 | 10 | 6233 | XP_011517647.1 | |
| XM_017015800.1 | 9 | 5738 | XP_016871289.1 | |
| XM_017015802.1 | 11 | 8116 | XP_016871291.1 | |
| XM_017015803.1 | 12 | 5730 | XP_016871292.1 | |
| XM_017015804.1 | 12 | 5568 | XP_016871293.1 | |
| XM_017015805.1 | 11 | 5765 | XP_016871294.1 | |
| XM_017015806.1 | 12 | 5707 | XP_016871295.1 | |
| XM_017015809.1 | 8 | 5682 | XP_016871298.1 | |
| XM_017015811.1 | 9 | 5628 | XP_016871300.1 | |
| XM_017015812.1 | 9 | 5422 | XP_016871301.1 | |
| BX537961.1 | 12 | 5722 | CAD97925.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831801 | Chr.10:17085685 - 17265571 on Build GRCh38 | Gain |
|
| nsv1039697 | Chr.10:17006669 - 17549229 on Build GRCh38 | Loss |
|
| nsv1045115 | Chr.10:17037131 - 17167193 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map