Assay Details
Target Gene Details
Entrez Gene ID: | 51170 |
Gene Name: | hydroxysteroid 17-beta dehydrogenase 11 |
Gene Aliases: |
17-BETA-HSD11, 17-BETA-HSDXI, 17BHSD11, DHRS8, PAN1B, RETSDR2, SDR16C2 |
Location: |
Chr.4:87336522-87391303 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HSD17B11 | NM_016245.4 | 7 | 1250 | NP_057329.3 |
| AF126780.1 | 7 | 1205 | AAF06939.1 | |
| AF273056.1 | 7 | 1103 | AAM44459.1 | |
| AK075348.1 | 7 | 1134 | BAC11560.1 | |
| AY358553.1 | 7 | 1117 | AAQ88917.1 | |
| BC008650.1 | 7 | 1140 | AAH08650.1 | |
| BC014327.1 | 7 | 1116 | AAH14327.1 | |
| BC016367.1 | 7 | 1147 | AAH16367.1 | |
| BC021673.1 | 7 | 1121 | AAH21673.1 | |
| BC036001.1 | 7 | 1141 | AAH36001.1 | |
| BM976691.1 | ||||
| CA775960.1 | 1 | 550 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1004518 | Chr.4:87297321 - 87433251 on Build GRCh38 | Gain |
|
| esv3601271 | Chr.4:87289406 - 87409657 on Build GRCh38 | Loss |
|
| nsv4412 | Chr.4:87316661 - 87362216 on Build GRCh38 | Deletion |
|
| nsv1002789 | Chr.4:87318188 - 87685333 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map