Assay Details
Target Gene Details
Entrez Gene ID: | 25879 |
Gene Name: | DDB1 and CUL4 associated factor 13 |
Gene Aliases: |
GM83, HSPC064, WDSOF1 |
Location: |
Chr.8:103414714-103443453 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DCAF13 | NM_015420.6 | 1 | 621 | NP_056235.4 |
| AK027799.1 | 1 | 622 | BAB55377.1 | |
| AK074725.1 | 1 | 586 | BAC11163.1 | |
| BC101810.1 | 1 | 411 | AAI01811.1 | |
| BC112042.1 | 1 | 411 | AAI12043.1 |
Target Gene Details
Entrez Gene ID: | 81034 |
Gene Name: | solute carrier family 25 member 32 |
Gene Aliases: |
MFT, MFTC, RREI |
Location: |
Chr.8:103398638-103415335 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC25A32 | NM_030780.4 | NP_110407.2 | ||
| NR_102337.1 | ||||
| NR_102338.1 | ||||
| DB087013.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv611832 | Chr.8:103388580 - 103469507 on Build GRCh38 | Loss |
|
| dgv7279n100 | Chr.8:103349626 - 103867456 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map