Assay Details
Target Gene Details
Entrez Gene ID: | 51112 |
Gene Name: | trafficking protein particle complex 12 |
Gene Aliases: |
CGI-87, TTC-15, TTC15 |
Location: |
Chr.2:3379245-3479571 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TRAPPC12 | NM_001321102.1 | 2 | 1036 | NP_001308031.1 |
| NM_016030.5 | 2 | 1073 | NP_057114.5 | |
| XM_011510350.2 | 2 | 1503 | XP_011508652.1 | |
| XM_011510352.2 | 2 | 1503 | XP_011508654.1 | |
| XM_011510353.2 | 2 | 1503 | XP_011508655.1 | |
| XM_011510354.2 | 2 | 1503 | XP_011508656.1 | |
| XM_017004257.1 | 2 | 1503 | XP_016859746.1 | |
| AK094181.1 | 3 | 1023 | BAC04301.1 | |
| AK094242.1 | 1 | 447 | ||
| AK098327.1 | 2 | 1038 | ||
| BC014164.2 | 2 | 950 | AAH14164.2 | |
| BC017475.1 | 1 | 71 | AAH17475.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1011656 | Chr.2:2931099 - 3576389 on Build GRCh38 | Gain |
|
| nsv1003355 | Chr.2:2560088 - 3543298 on Build GRCh38 | Gain |
|
| nsv580773 | Chr.2:3376797 - 3390663 on Build GRCh38 | Loss |
|
| dgv3702n100 | Chr.2:3361936 - 3733306 on Build GRCh38 | Gain |
|
| esv3589655 | Chr.2:3368551 - 3433441 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map