Assay Details
Target Gene Details
Entrez Gene ID: | 114815 |
Gene Name: | sortilin related VPS10 domain containing receptor 1 |
Gene Aliases: |
hSorCS |
Location: |
Chr.10:106573663-107181138 on Build GRCh38 |
Assay Gene Location: | Within Exon 30 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SORCS1 | NM_001013031.2 | 27 | 6183 | NP_001013049.1 |
| NM_001206570.1 | 26 | 6022 | NP_001193499.1 | |
| NM_001206572.1 | 27 | 6453 | NP_001193501.1 | |
| NM_052918.4 | 26 | 6292 | NP_443150.3 | |
| XM_017015614.1 | XP_016871103.1 | |||
| XM_017015615.1 | XP_016871104.1 | |||
| XM_017015616.1 | XP_016871105.1 | |||
| XM_017015617.1 | XP_016871106.1 | |||
| XM_017015618.1 | XP_016871107.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831977 | Chr.10:106466255 - 106653420 on Build GRCh38 | Gain |
|
| nsv1047446 | Chr.10:105797401 - 106616713 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map