Assay Details
Target Gene Details
Entrez Gene ID: | 84678 |
Gene Name: | lysine demethylase 2B |
Gene Aliases: |
CXXC2, FBXL10, Fbl10, JHDM1B, PCCX2 |
Location: |
Chr.12:121402289-121582268 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 26 - Exon 26 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KDM2B | NM_001005366.1 | NP_001005366.1 | ||
| NM_032590.4 | NP_115979.3 | |||
| XM_011538867.2 | XP_011537169.1 | |||
| AA504464.1 | ||||
| AB031230.1 | BAA97672.1 | |||
| CA430876.1 |
More Information
Set Membership: |
|

Genomic Map