Assay Details
Target Gene Details
Entrez Gene ID: | 25766 |
Gene Name: | pre-mRNA processing factor 40 homolog B |
Gene Aliases: |
HYPC |
Location: |
Chr.12:49622717-49644669 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRPF40B | NM_001031698.2 | 8 | 675 | NP_001026868.2 |
| NM_012272.2 | 7 | 965 | NP_036404.1 | |
| XM_006719324.3 | 7 | 1729 | XP_006719387.1 | |
| XM_006719325.3 | 7 | 1729 | XP_006719388.1 | |
| XM_011538135.2 | 7 | 1729 | XP_011536437.1 | |
| XM_011538136.2 | 7 | 1729 | XP_011536438.1 | |
| XM_011538137.2 | 6 | 1648 | XP_011536439.1 | |
| XM_011538138.2 | 6 | 1618 | XP_011536440.1 | |
| XM_011538139.2 | 8 | 669 | XP_011536441.1 | |
| XM_011538140.2 | 8 | 669 | XP_011536442.1 | |
| XM_011538141.1 | 7 | 495 | XP_011536443.1 | |
| XM_011538143.2 | 7 | 511 | XP_011536445.1 | |
| XM_011538144.2 | 6 | 384 | XP_011536446.1 | |
| XM_017019135.1 | 7 | 432 | XP_016874624.1 | |
| XM_017019136.1 | 6 | 384 | XP_016874625.1 | |
| XM_017019137.1 | 6 | 384 | XP_016874626.1 | |
| XM_017019138.1 | 7 | 1729 | XP_016874627.1 | |
| XM_017019139.1 | 3 | 395 | XP_016874628.1 | |
| AF049525.1 | 6 | 329 | AAC27503.1 | |
| AK123353.1 | 5 | 1339 | ||
| AL834216.1 | 7 | 965 | CAD38898.1 | |
| BC050398.2 | 8 | 511 | AAH50398.1 | |
| BC067364.1 | 7 | 496 | AAH67364.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832406 | Chr.12:49466910 - 49668353 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map