Assay Details
Target Gene Details
Entrez Gene ID: | 25953 |
Gene Name: | paroxysmal nonkinesigenic dyskinesia |
Gene Aliases: |
BRP17, DYT8, FKSG19, FPD1, KIPP1184, MR-1, MR1, PDC, PKND1, TAHCCP2 |
Location: |
Chr.2:218270392-218346793 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PNKD | NM_001077399.2 | NP_001070867.1 | ||
| NM_015488.4 | NP_056303.3 | |||
| XM_017003771.1 | XP_016859260.1 | |||
| AB033010.1 | BAA86498.1 | |||
| AF318057.1 | AAL25716.1 | |||
| AF417001.1 | AAL08573.1 | |||
| AF418285.1 | AAL08580.1 | |||
| AK289867.1 | ||||
| AL080092.1 | CAB45707.2 | |||
| BC021118.1 | AAH21118.1 | |||
| BC036457.2 | AAH36457.1 | |||
| BQ028777.1 |
More Information
Set Membership: |
|

Genomic Map