Assay Details
Target Gene Details
Entrez Gene ID: | 64376 |
Gene Name: | IKAROS family zinc finger 5 |
Gene Aliases: |
PEGASUS, ZNFN1A5 |
Location: |
Chr.10:122990806-123008850 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IKZF5 | NM_001271840.1 | 5 | 1786 | NP_001258769.1 |
| NR_073475.1 | 5 | 1830 | ||
| NR_073476.1 | 5 | 1826 | ||
| NR_073477.1 | 5 | 1817 | ||
| NR_073478.1 | 4 | 1679 | ||
| NR_073479.1 | 6 | 1938 | ||
| XM_006717947.3 | 6 | 1900 | XP_006718010.1 | |
| XM_017016550.1 | 5 | 1778 | XP_016872039.1 | |
| XM_017016551.1 | 6 | 1874 | XP_016872040.1 | |
| XM_017016552.1 | 5 | 1766 | XP_016872041.1 | |
| XM_017016553.1 | 5 | 2275 | XP_016872042.1 | |
| XM_017016554.1 | 6 | 2383 | XP_016872043.1 | |
| XM_017016555.1 | 5 | 2262 | XP_016872044.1 | |
| AK226143.1 | 5 | 1767 | ||
| BX647958.1 | 5 | 1779 | CAH56199.1 | |
| CR749800.1 | 5 | 1772 | CAH18660.1 |
Target Gene Details
Entrez Gene ID: | 118672 |
Gene Name: | phosphoseryl-tRNA kinase |
Gene Aliases: |
C10orf89 |
Location: |
Chr.10:122980040-122997519 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PSTK | AK094271.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv7586 | Chr.10:122982687 - 123016052 on Build GRCh38 | Insertion |
|
| nsv832010 | Chr.10:122826964 - 123037874 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map