Assay Details
Target Gene Details
Entrez Gene ID: | 6774 |
Gene Name: | signal transducer and activator of transcription 3 |
Gene Aliases: |
ADMIO, ADMIO1, APRF, HIES |
Location: |
Chr.17:42313324-42388505 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 24 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| STAT3 | XM_011525145.2 | XP_011523447.1 | ||
| XM_011525146.2 | XP_011523448.1 | |||
| XM_017024973.1 | XP_016880462.1 | |||
| AK024535.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833449 | Chr.17:42274854 - 42421081 on Build GRCh38 | Loss |
|
| nsv1056904 | Chr.17:42025823 - 42363204 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map