Assay Details
Target Gene Details
Entrez Gene ID: | 26135 |
Gene Name: | SERPINE1 mRNA binding protein 1 |
Gene Aliases: |
CGI-55, CHD3IP, HABP4L, PAI-RBP1, PAIRBP1 |
Location: |
Chr.1:67407810-67430440 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SERBP1 | NM_001018067.1 | 8 | 5661 | NP_001018077.1 |
| NM_001018068.1 | 8 | 5643 | NP_001018078.1 | |
| NM_001018069.1 | 8 | 5616 | NP_001018079.1 | |
| NM_015640.3 | 8 | 5598 | NP_056455.3 | |
| AK095960.1 | 1 | 2038 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv461773 | Chr.1:67356694 - 67442961 on Build GRCh38 | Loss |
|
| nsv1254 | Chr.1:67394481 - 67428346 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map