Assay Details
Target Gene Details
Entrez Gene ID: | 2137 |
Gene Name: | exostosin like glycosyltransferase 3 |
Gene Aliases: |
BOTV, EXTL1L, EXTR1, REGR, RPR |
Location: |
Chr.8:28701473-28753690 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EXTL3 | NM_001440.3 | 3 | 430 | NP_001431.1 |
| NR_073468.1 | ||||
| NR_073469.1 | 3 | 430 | ||
| XM_011544440.2 | 2 | 206 | XP_011542742.1 | |
| AB007042.1 | 3 | 267 | BAA24080.1 | |
| AB011091.1 | 3 | 289 | ||
| AF001690.1 | 3 | 275 | AAC39598.1 | |
| AF083551.1 | 3 | 267 | AAD42041.1 | |
| BI911162.1 | 3 | 298 | ||
| DA222815.1 | ||||
| DA757362.1 |
More Information
Set Membership: |
|

Genomic Map