Assay Details
Target Gene Details
Entrez Gene ID: | 10809 |
Gene Name: | StAR related lipid transfer domain containing 10 |
Gene Aliases: |
CGI-52, NY-CO-28, PCTP2, SDCCAG28 |
Location: |
Chr.11:72754729-72793705 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| STARD10 | NM_006645.2 | 7 | 1700 | NP_006636.2 |
| AF151810.1 | 7 | 1236 | AAD34047.1 | |
| AK095668.1 | 7 | 1616 | ||
| AK315255.1 | 8 | 1494 | ||
| BC007919.2 | 8 | 1219 | AAH07919.1 | |
| BC014033.2 | 8 | 1219 | AAH14033.1 | |
| JX424571.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832207 | Chr.11:72561706 - 72758077 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map