Assay Details
Target Gene Details
Entrez Gene ID: | 23764 |
Gene Name: | MAF bZIP transcription factor F |
Gene Aliases: |
U-MAF, hMafF |
Location: |
Chr.22:38201932-38216511 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAFF | NM_001161572.1 | 3 | 560 | NP_001155044.1 |
| NM_001161573.1 | 3 | 318 | NP_001155045.1 | |
| NM_001161574.1 | 2 | 467 | NP_001155046.1 | |
| NM_012323.3 | 3 | 534 | NP_036455.1 | |
| AB025247.1 | 3 | 368 | BAA86871.1 | |
| AJ010857.1 | 3 | 455 | CAB52435.1 | |
| AK002001.1 | 3 | 475 | BAA92029.1 | |
| AK097953.1 | 3 | 425 | ||
| AK300932.1 | 2 | 404 | ||
| BC015037.1 | 3 | 407 | AAH15037.1 | |
| BC067751.1 | 3 | 474 | AAH67751.1 | |
| DA209378.1 | ||||
| DA425613.1 | 3 | 318 | ||
| DA654696.1 | 3 | 502 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834194 | Chr.22:38063495 - 38240504 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map