Assay Details
Target Gene Details
Entrez Gene ID: | 2676 |
Gene Name: | GDNF family receptor alpha 3 |
Gene Aliases: |
GDNFR3 |
Location: |
Chr.5:138252380-138274564 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GFRA3 | NM_001496.3 | 8 | 1550 | NP_001487.2 |
| AF086292.1 | 1 | 132 | ||
| AK297693.1 | 7 | 1411 | ||
| AW020606.2 | 1 | 81 | ||
| AY358997.1 | 8 | 1447 | AAQ89356.1 | |
| AY359037.1 | 7 | 1458 | AAQ89396.1 | |
| BC037951.1 | 8 | 1435 | AAH37951.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3389910 | Chr.5:138252164 - 138254262 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map