Assay Details
Target Gene Details
Entrez Gene ID: | 5104 |
Gene Name: | serpin family A member 5 |
Gene Aliases: |
PAI-3, PAI3, PCI, PCI-B, PLANH3, PROCI |
Location: |
Chr.14:94581369-94593120 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SERPINA5 | NM_000624.5 | NP_000615.3 | ||
| AI114432.1 | ||||
| AK096131.1 | ||||
| AK298279.1 | ||||
| AL080185.1 | CAB45766.1 | |||
| BC008915.2 | AAH08915.1 | |||
| J02639.1 | AAA35688.1 | |||
| S58545.1 | AAB26244.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv470662 | Chr.14:94296542 - 94798793 on Build GRCh38 | Gain |
|
| nsv565625 | Chr.14:94567019 - 94603850 on Build GRCh38 | Loss |
|
| nsv565623 | Chr.14:94393116 - 94635006 on Build GRCh38 | Gain |
|
| nsv832865 | Chr.14:94499457 - 94667894 on Build GRCh38 | Gain |
|
| esv3635384 | Chr.14:94569665 - 94707857 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map