Assay Details
Target Gene Details
Entrez Gene ID: | 26137 |
Gene Name: | zinc finger and BTB domain containing 20 |
Gene Aliases: |
DPZF, HOF, ODA-8S, PRIMS, ZNF288 |
Location: |
Chr.3:114314500-115147280 on Build GRCh38 |
Assay Gene Location: | Within Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZBTB20 | NM_001164342.2 | 5 | 2369 | NP_001157814.1 |
| NM_001164343.2 | 12 | 2831 | NP_001157815.1 | |
| NM_001164344.2 | 8 | 2795 | NP_001157816.1 | |
| NM_001164345.2 | 7 | 2749 | NP_001157817.1 | |
| NM_001164346.2 | 6 | 2388 | NP_001157818.1 | |
| NM_001164347.2 | 6 | 2410 | NP_001157819.1 | |
| NM_015642.5 | 10 | 2593 | NP_056457.3 | |
| NR_121662.1 | 7 | 851 | ||
| AF139460.2 | 5 | 2375 | AAG28340.2 | |
| AK129968.1 | 1 | 912 | ||
| AK225788.1 | 7 | 2745 | ||
| AK290116.1 | 3 | 2131 | ||
| AK315307.1 | 6 | 2388 | ||
| AL050276.1 | 8 | 2458 | CAB43377.1 | |
| BC029041.1 | 6 | 2410 | AAH29041.1 | |
| BX647778.1 | 10 | 2594 | CAH56190.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv829679 | Chr.3:114331081 - 114533888 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map