Assay Details
Target Gene Details
Entrez Gene ID: | 53938 |
Gene Name: | peptidylprolyl isomerase like 3 |
Gene Aliases: |
CYPJ |
Location: |
Chr.2:200870907-200889333 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PPIL3 | NM_032472.3 | 7 | 831 | NP_115861.1 |
| NM_130906.2 | 7 | 819 | NP_570981.1 | |
| XM_005246651.3 | 7 | 750 | XP_005246708.1 | |
| XM_005246652.4 | 7 | 738 | XP_005246709.1 | |
| XM_011511358.2 | 7 | 924 | XP_011509660.1 | |
| XM_017004352.1 | 7 | 941 | XP_016859841.1 | |
| XM_017004353.1 | 7 | 680 | XP_016859842.1 | |
| XM_017004354.1 | 7 | 1114 | XP_016859843.1 | |
| XM_017004355.1 | 7 | 1138 | XP_016859844.1 | |
| XM_017004356.1 | 7 | 1102 | XP_016859845.1 | |
| XM_017004357.1 | 8 | 1021 | XP_016859846.1 | |
| XM_017004358.1 | 8 | 844 | XP_016859847.1 | |
| XM_017004359.1 | 8 | 1028 | XP_016859848.1 | |
| XM_017004360.1 | 6 | 841 | XP_016859849.1 | |
| AF146799.1 | 7 | 688 | AAO64723.1 | |
| AF251049.1 | 7 | 831 | AAK34939.1 | |
| AF271652.1 | 7 | 754 | AAG44766.1 | |
| AF414127.1 | 6 | 755 | AAP97286.1 | |
| AK297804.1 | ||||
| AY257468.1 | 6 | 875 | AAP13527.1 | |
| BC007693.2 | 7 | 1260 | AAH07693.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs150825161] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map