Assay Details
Target Gene Details
Entrez Gene ID: | 4734 |
Gene Name: | neural precursor cell expressed, developmentally down-regulated 4, E3 ubiquitin protein ligase |
Gene Aliases: |
NEDD4-1, RPF1 |
Location: |
Chr.15:55826917-55993746 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 28 - Exon 29 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NEDD4 | NM_001284338.1 | NP_001271267.1 | ||
| NM_001284339.1 | NP_001271268.1 | |||
| NM_001284340.1 | NP_001271269.1 | |||
| NM_006154.3 | NP_006145.2 | |||
| NM_198400.3 | NP_940682.2 | |||
| NR_104302.1 | ||||
| XM_011521624.2 | XP_011519926.1 | |||
| XM_011521625.2 | XP_011519927.1 | |||
| XM_011521626.1 | XP_011519928.1 | |||
| XM_011521627.1 | XP_011519929.1 | |||
| AK304394.1 | ||||
| AK307541.1 | ||||
| AL832063.1 | ||||
| AL832359.1 | ||||
| AL833293.1 | ||||
| AL833300.1 | ||||
| AY550969.1 | ||||
| BC136605.1 | ||||
| BC144284.1 | ||||
| BC144285.1 | ||||
| BC152452.1 | ||||
| BC152562.1 | ||||
| D42055.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833017 | Chr.15:55673759 - 55865714 on Build GRCh38 | Loss |
|
| nsv569539 | Chr.15:55430015 - 55963576 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map