Assay Details
Target Gene Details
Entrez Gene ID: | 10961 |
Gene Name: | endoplasmic reticulum protein 29 |
Gene Aliases: |
C12orf8, ERp28, ERp31, HEL-S-107, PDI-DB, PDIA9 |
Location: |
Chr.12:112013340-112023220 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ERP29 | NM_001034025.1 | 2 | 1013 | NP_001029197.1 |
| NM_006817.3 | 3 | 1152 | NP_006808.1 | |
| XM_017018720.1 | 2 | 1021 | XP_016874209.1 | |
| BE794579.1 |
More Information
Set Membership: |
|

Genomic Map