Assay Details
Target Gene Details
Entrez Gene ID: | 23484 |
Gene Name: | leptin receptor overlapping transcript-like 1 |
Gene Aliases: |
HSPC112, Vps55, my047 |
Location: |
Chr.8:30095406-30137706 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LEPROTL1 | NM_001128208.1 | NP_001121680.1 | ||
| NM_015344.2 | 4 | 1866 | NP_056159.2 | |
| AF063605.1 | 4 | 1843 | AAC98697.1 | |
| AK075363.1 | 4 | 1810 | ||
| AY358674.1 | 4 | 1838 | AAQ89037.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv6138 | Chr.8:30084206 - 30118638 on Build GRCh38 | Insertion |
|
| nsv831277 | Chr.8:29977428 - 30180297 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map