Assay Details
Target Gene Details
Entrez Gene ID: | 163702 |
Gene Name: | interferon lambda receptor 1 |
Gene Aliases: |
CRF2/12, IFNLR, IL-28R1, IL28RA, LICR2 |
Location: |
Chr.1:24154156-24187275 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IFNLR1 | NM_170743.3 | 7 | 3666 | NP_734464.1 |
| NM_173064.2 | 7 | 3579 | NP_775087.1 | |
| NM_173065.2 | 6 | 3535 | NP_775088.1 | |
| XM_006710394.3 | 6 | 3730 | XP_006710457.2 | |
| XM_017000479.1 | 6 | 3695 | XP_016855968.1 | |
| AF439325.1 | 7 | 3639 | AAN63632.1 | |
| AK160364.1 | 7 | 3642 | ||
| BC140872.1 | 7 | 3715 |
More Information
Set Membership: |
|

Genomic Map