Assay Details
Target Gene Details
Entrez Gene ID: | 6401 |
Gene Name: | selectin E |
Gene Aliases: |
CD62E, ELAM, ELAM1, ESEL, LECAM2 |
Location: |
Chr.1:169722640-169734079 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SELE | NM_000450.2 | 14 | 3313 | NP_000441.2 |
| BC142677.1 | 14 | 3300 | ||
| BC142711.1 | 14 | 3300 | ||
| M24736.1 | 14 | 3272 | AAA52376.1 | |
| M30640.1 | 14 | 3296 | AAA52377.1 |
More Information
Set Membership: |
|

Genomic Map