Assay Details
Target Gene Details
Entrez Gene ID: | 91851 |
Gene Name: | chordin like 1 |
Gene Aliases: |
CHL, MGC1, MGCN, NRLN1, VOPT, dA141H5.1 |
Location: |
Chr.X:110673856-110796058 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CHRDL1 | NM_001143981.1 | 12 | 2026 | NP_001137453.1 |
| NM_001143982.1 | 12 | 2023 | NP_001137454.1 | |
| NM_001143983.2 | 10 | 1786 | NP_001137455.2 | |
| NM_145234.3 | 12 | 1868 | NP_660277.2 | |
| XM_005262221.1 | 12 | 1843 | XP_005262278.1 | |
| XM_005262222.3 | 12 | 1840 | XP_005262279.1 | |
| XM_005262223.1 | 12 | 1805 | XP_005262280.1 | |
| XM_005262224.1 | 12 | 1824 | XP_005262281.1 | |
| XM_017029959.1 | 12 | 1828 | XP_016885448.1 | |
| AK092245.1 | 9 | 1453 | ||
| AK293106.1 | 12 | 1773 | ||
| AK297563.1 | ||||
| BC002909.2 | 12 | 1785 | AAH02909.2 |
More Information
Set Membership: |
|

Genomic Map