Assay Details
Target Gene Details
Entrez Gene ID: | 11269 |
Gene Name: | DEAD-box helicase 19B |
Gene Aliases: |
DBP5, DDX19, RNAh |
Location: |
Chr.16:70289767-70335213 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DDX19B | NM_001014449.2 | NP_001014449.1 | ||
| NM_001014451.2 | NP_001014451.1 | |||
| NM_001257172.1 | NP_001244101.1 | |||
| NM_001257173.1 | NP_001244102.1 | |||
| NM_001257174.1 | NP_001244103.1 | |||
| NM_001257175.1 | NP_001244104.1 | |||
| NM_007242.5 | NP_009173.1 | |||
| XM_006721127.2 | 11 | 1778 | XP_006721190.1 | |
| XM_011522832.2 | 13 | 1912 | XP_011521134.1 | |
| XM_011522833.2 | 12 | 1819 | XP_011521135.1 | |
| XM_011522834.2 | 12 | 1801 | XP_011521136.1 | |
| XM_017022890.1 | 12 | 1894 | XP_016878379.1 | |
| AF353720.1 | AAK40102.1 | |||
| AK027378.1 | ||||
| AK223115.1 | BAD96835.1 | |||
| AK296868.1 | 9 | 1657 | ||
| AK316346.1 | ||||
| AL136639.1 | CAB66574.1 | |||
| BC003626.2 | AAH03626.1 | |||
| BC010008.2 | AAH10008.1 | |||
| BC110805.1 | AAI10806.1 |
Target Gene Details
Entrez Gene ID: | 100506083 |
Gene Name: | uncharacterized LOC100506083 |
Gene Aliases: |
- |
Location: |
Chr.16:70315640-70346747 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC100506083 | NR_039997.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2714670 | Chr.16:70331662 - 70370229 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map