Assay Details
Target Gene Details
Entrez Gene ID: | 83892 |
Gene Name: | potassium channel tetramerization domain containing 10 |
Gene Aliases: |
BTBD28, MSTP028, ULRO61, hBACURD3 |
Location: |
Chr.12:109448655-109477544 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KCTD10 | NM_001317395.1 | 7 | 3990 | NP_001304324.1 |
| NM_001317399.1 | 7 | 3918 | NP_001304328.1 | |
| NM_031954.4 | 7 | 3987 | NP_114160.1 | |
| NR_133897.1 | 6 | 3817 | ||
| NR_133898.1 | 6 | 3773 | ||
| AY205299.1 | 7 | 3793 | AAO47716.1 | |
| BC040062.1 | 7 | 3740 | AAH40062.1 |
More Information
Set Membership: |
|

Genomic Map