Assay Details
Target Gene Details
Entrez Gene ID: | 54928 |
Gene Name: | inositol monophosphatase domain containing 1 |
Gene Aliases: |
GPAPP, IMP 3, IMP-3, IMPA3 |
Location: |
Chr.8:56957929-56993874 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IMPAD1 | NM_017813.4 | 5 | 1644 | NP_060283.3 |
| AK000428.1 | BAA91158.1 | |||
| BC017797.1 | AAH17797.1 | |||
| BC067814.1 | 5 | 1617 | AAH67814.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3891400 | Chr.8:56933474 - 57202184 on Build GRCh38 | Gain |
|
| nsv1029443 | Chr.8:56835555 - 57145097 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map