Assay Details
Target Gene Details
Entrez Gene ID: | 3703 |
Gene Name: | STT3A, catalytic subunit of the oligosaccharyltransferase complex |
Gene Aliases: |
ITM1, STT3-A, TMC |
Location: |
Chr.11:125592795-125622759 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| STT3A | NM_001278503.1 | 8 | 816 | NP_001265432.1 |
| NM_001278504.1 | 6 | 605 | NP_001265433.1 | |
| NM_152713.4 | 7 | 728 | NP_689926.1 | |
| XM_011542807.2 | 7 | 755 | XP_011541109.1 | |
| AK290040.1 | 7 | 678 | ||
| AK290657.1 | 7 | 676 | ||
| AK295892.1 | 6 | 533 | ||
| AK308902.1 | 7 | 671 | ||
| BC020965.2 | 7 | 661 | AAH20965.1 | |
| BC048348.1 | 6 | 614 | ||
| BT007100.1 | 6 | 569 | AAP35764.1 | |
| BX649102.1 | 8 | 754 | ||
| L38961.1 | 7 | 675 | AAB05994.1 | |
| L47337.1 | 7 | 685 | AAL77539.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv32964 | Chr.11:125495069 - 126046387 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map