Assay Details
Target Gene Details
Entrez Gene ID: | 56853 |
Gene Name: | CUGBP, Elav-like family member 4 |
Gene Aliases: |
BRUNOL4, CELF-4 |
Location: |
Chr.18:37243040-37566052 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
Target Gene Details
Entrez Gene ID: | 105372068 |
Gene Name: | uncharacterized LOC105372068 |
Gene Aliases: |
- |
Location: |
Chr.18:37274460-37276400 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC105372068 | NR_134588.1 | |||
| AI200809.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833621 | Chr.18:37127725 - 37302713 on Build GRCh38 | Loss |
|
| nsv953532 | Chr.18:37246238 - 37292337 on Build GRCh38 | Deletion |
|
| nsv576654 | Chr.18:37249923 - 37280895 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map