Assay Details
Target Gene Details
Entrez Gene ID: | 55720 |
Gene Name: | TSR1, ribosome maturation factor |
Gene Aliases: |
- |
Location: |
Chr.17:2322678-2337384 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TSR1 | NM_018128.4 | NP_060598.3 | ||
| AB037822.1 | ||||
| AK001396.1 | BAA91667.1 | |||
| AK026565.1 | ||||
| BC019090.2 | AAH19090.2 | |||
| BC110851.1 | AAI10852.1 | |||
| BC126110.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv574224 | Chr.17:2269769 - 2423186 on Build GRCh38 | Gain |
|
| nsv1067416 | Chr.17:2319186 - 2343258 on Build GRCh38 | Loss |
|
| nsv469765 | Chr.17:2213155 - 2400446 on Build GRCh38 | Loss |
|
| nsv833336 | Chr.17:2176922 - 2400456 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map